accessibility menu, dialog, popup

UserName

CAS RN 930-69-8

NaS

CAS RN 930-69-8

IUPAC Name: sodium phenylsulfanide
Synonyms: sodium phenylsulfanide
Molecular Weight (g/mol): 132.16
Molecular Formula: C6H5NaS
InChi Key: RZWQDAUIUBVCDD-UHFFFAOYSA-M
SMILES: [Na+].[S-]C1=CC=CC=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results