accessibility menu, dialog, popup

UserName

CAS RN 930111-10-7

CH3H3CCH3OONOBr

CAS RN 930111-10-7

IUPAC Name: tert-butyl 4-[(4-bromophenyl)methoxy]piperidine-1-carboxylate
Synonyms: tert-butyl 4-[(4-bromophenyl)methoxy]piperidine-1-carboxylate
Molecular Weight (g/mol): 370.29
Molecular Formula: C17H24BrNO3
InChi Key: HGSFDBSIEHXEQD-UHFFFAOYSA-N
SMILES: CC(C)(C)OC(=O)N1CCC(CC1)OCC1=CC=C(Br)C=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results