accessibility menu, dialog, popup

UserName

H3CNNBrHBr

CAS RN 934570-40-8

IUPAC Name: 2-(bromomethyl)-1-methyl-1H-1,3-benzodiazole hydrobromide
Synonyms: 2-(bromomethyl)-1-methyl-1,3-benzodiazole hydrobromide
Molecular Weight (g/mol): 306
Molecular Formula: C9H10Br2N2
InChi Key: WHMCWOHHFOGLGP-UHFFFAOYSA-N
SMILES: Br.CN1C(CBr)=NC2=CC=CC=C12

 No Refinements