accessibility menu, dialog, popup

UserName

HOOFFFHOFFF

CAS RN 936107-87-8

IUPAC Name: 3,3,3-trifluoro-1-(3,3,3-trifluoro-1-hydroxypropoxy)propan-1-ol
Synonyms: 3,3,3-trifluoro-1-(3,3,3-trifluoro-1-hydroxypropoxy)propan-1-ol
Molecular Weight (g/mol): 242.12
Molecular Formula: C6H8F6O3
InChi Key: MBQFJCIIFZJMTG-UHFFFAOYNA-N
SMILES: OC(CC(F)(F)F)OC(O)CC(F)(F)F

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results