accessibility menu, dialog, popup

UserName

H3CH3CCH3O

CAS RN 938-16-9

IUPAC Name: 2,2-dimethyl-1-phenylpropan-1-one
Synonyms: 2,2-dimethylpropiophenone
Molecular Weight (g/mol): 162.23
Molecular Formula: C11H14O
InChi Key: OECPUBRNDKXFDX-UHFFFAOYSA-N
SMILES: CC(C)(C)C(=O)C1=CC=CC=C1
Boiling Point 
  • (3)
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results