accessibility menu, dialog, popup

UserName

CAS RN 93962-84-6

H2COO

CAS RN 93962-84-6

IUPAC Name: {tricyclo[5.2.1.0²,⁶]decan-3-yl}methyl prop-2-enoate
Synonyms: tricyclo[5.2.1.0²,⁶]decan-3-ylmethyl prop-2-enoate
Molecular Weight (g/mol): 220.31
Molecular Formula: C14H20O2
InChi Key: PMPNHSZLJPXGCD-UHFFFAOYNA-N
SMILES: C=CC(=O)OCC1CCC2C3CCC(C3)C12
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results