accessibility menu, dialog, popup

UserName

BOOH3CCH3H3CH3CFNH2F

CAS RN 939968-08-8

IUPAC Name: 2,6-difluoro-4-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)aniline
Synonyms: 2,6-difluoro-4-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)aniline
Molecular Weight (g/mol): 255.07
Molecular Formula: C12H16BF2NO2
InChi Key: JGOZEXIYNJERIP-UHFFFAOYSA-N
SMILES: CC1(C)OB(OC1(C)C)C1=CC(F)=C(N)C(F)=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results