accessibility menu, dialog, popup

UserName

CAS RN 941717-01-7

CH3NSBr

CAS RN 941717-01-7

IUPAC Name: 4-(2-bromophenyl)-2-methyl-1,3-thiazole
Synonyms: 4-(2-bromophenyl)-2-methyl-1,3-thiazole
Molecular Weight (g/mol): 254.15
Molecular Formula: C10H8BrNS
InChi Key: SXJQVZDDCODIST-UHFFFAOYSA-N
SMILES: CC1=NC(=CS1)C1=CC=CC=C1Br

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results