accessibility menu, dialog, popup

UserName

NFNCl

CAS RN 942206-09-9

IUPAC Name: 2-chloro-2'-fluoro-3,3'-bipyridine
Synonyms: 2-chloro-2'-fluoro-3,3'-bipyridine
Molecular Weight (g/mol): 208.62
Molecular Formula: C10H6ClFN2
InChi Key: XJKOECSGYQHCPP-UHFFFAOYSA-N
SMILES: FC1=C(C=CC=N1)C1=C(Cl)N=CC=C1
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results