accessibility menu, dialog, popup

UserName

CAS RN 942206-23-7

NIClCl

CAS RN 942206-23-7

IUPAC Name: 2,5-dichloro-3-iodopyridine
Synonyms: 2,5-dichloro-3-iodopyridine
Molecular Weight (g/mol): 273.88
Molecular Formula: C5H2Cl2IN
InChi Key: VYVNSDZZIATDSY-UHFFFAOYSA-N
SMILES: ClC1=CC(I)=C(Cl)N=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results