CAS RN 94421-68-8
IUPAC Name:
(5Z,8Z,11Z,14Z)-N-(2-hydroxyethyl)icosa-5,8,11,14-tetraenamide
Synonyms:
anandamide n-6) anandamide (20.4 AEA n-6) anandamide(20:4 n-arachidonoylethanolamide anandamide (20.4,n-6) anandamide(20:4, n-6) arachidonoylethanolamide arachidonoyl ethanolamide n arachidonoyl 2 hydroxyethylamide
Molecular Weight (g/mol):
347.54
Molecular Formula:
C22H37NO2
InChi Key:
LGEQQWMQCRIYKG-DOFZRALJSA-N
SMILES:
CCCCC\C=C/C\C=C/C\C=C/C\C=C/CCCC(=O)NCCO