accessibility menu, dialog, popup

UserName

NNBrOOH

CAS RN 944896-42-8

IUPAC Name: 6-bromoimidazo[1,2-a]pyridine-3-carboxylic acid
Synonyms: 6-bromoimidazo[1,2-a]pyridine-3-carboxylic acid
Molecular Weight (g/mol): 241.04
Molecular Formula: C8H5BrN2O2
InChi Key: SDKJLYHPCBMDAD-UHFFFAOYSA-N
SMILES: OC(=O)C1=CN=C2C=CC(Br)=CN12

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results