accessibility menu, dialog, popup

UserName

CH3H3CCH3OONNNHNH3C

CAS RN 946409-15-0

IUPAC Name: tert-butyl 4-{4-[(methylamino)methyl]pyridin-2-yl}piperazine-1-carboxylate
Synonyms: tert-butyl 4-{4-[(methylamino)methyl]pyridin-2-yl}piperazine-1-carboxylate
Molecular Weight (g/mol): 306.41
Molecular Formula: C16H26N4O2
InChi Key: BNLNDKXJNWJYKP-UHFFFAOYSA-N
SMILES: CNCC1=CC(=NC=C1)N1CCN(CC1)C(=O)OC(C)(C)C

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results