accessibility menu, dialog, popup

UserName

CH3HNS

CAS RN 950603-22-2

IUPAC Name: methyl[(thian-4-yl)methyl]amine
Synonyms: methyl(thian-4-ylmethyl)amine
Molecular Weight (g/mol): 145.26
Molecular Formula: C7H15NS
InChi Key: FPKJBESBCAZWPI-UHFFFAOYSA-N
SMILES: CNCC1CCSCC1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results