CAS RN 950670-18-5
IUPAC Name:
3-bromo-5-fluoropyridine-2-carbonitrile
Synonyms:
3-bromo-5-fluoropyridine-2-carbonitrile
Molecular Weight (g/mol):
201
Molecular Formula:
C6H2BrFN2
InChi Key:
OLOSREGULYFXDD-UHFFFAOYSA-N
SMILES:
FC1=CC(Br)=C(N=C1)C#N
1
–
1
of
1
results
1
3-Bromo-5-fluoro-2-cyanopyridine, 97%
CAS: 950670-18-5 Molecular Formula: C6H2BrFN2 Molecular Weight (g/mol): 201 InChI Key: OLOSREGULYFXDD-UHFFFAOYSA-N Synonym: 3-bromo-5-fluoropicolinonitrile,3-bromo-5-fluoro-2-cyanopyridine,3-bromo-2-cyano-5-fluoropyridine,3-bromo-5-fluoro-2-pyridinecarbonitrile,abbypharma ap-18-5887,3-aromo-5-fluoropicolinonitrile PubChem CID: 46941477 IUPAC Name: 3-bromo-5-fluoropyridine-2-carbonitrile SMILES: C1=C(C=NC(=C1Br)C#N)F