accessibility menu, dialog, popup

UserName

CH3NOOBrNOO

CAS RN 95192-64-6

IUPAC Name: 5-bromo-2-methyl-1,3-dinitrobenzene
Synonyms: 5-bromo-2-methyl-1,3-dinitrobenzene
Molecular Weight (g/mol): 261.03
Molecular Formula: C7H5BrN2O4
InChi Key: UOGCLPDKGPPDHM-UHFFFAOYSA-N
SMILES: CC1=C(C=C(Br)C=C1[N+]([O-])=O)[N+]([O-])=O
Molecular Weight (g/mol) 
  • (2)
  • (2)
Quantity 
  • (2)
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)