accessibility menu, dialog, popup

UserName

FOClCl

CAS RN 95399-95-4

IUPAC Name: 2,3-dichloro-6-fluorobenzaldehyde
Synonyms: 2,3-dichloro-6-fluorobenzaldehyde
Molecular Weight (g/mol): 193
Molecular Formula: C7H3Cl2FO
InChi Key: BDPIHKOPKWGJCU-UHFFFAOYSA-N
SMILES: FC1=C(C=O)C(Cl)=C(Cl)C=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results