accessibility menu, dialog, popup

UserName

CH3H3CCH3OOHNSNH2N

CAS RN 956018-34-1

IUPAC Name: tert-butyl N-[2-(2-amino-1,3-thiazol-4-yl)ethyl]carbamate
Synonyms: tert-butyl N-[2-(2-amino-1,3-thiazol-4-yl)ethyl]carbamate
Molecular Weight (g/mol): 243.33
Molecular Formula: C10H17N3O2S
InChi Key: IIILZIJRVHFCGL-UHFFFAOYSA-N
SMILES: CC(C)(C)OC(=O)NCCC1=CSC(N)=N1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results