accessibility menu, dialog, popup

UserName

BONHHNSNH2OHHO

CAS RN 957060-76-3

IUPAC Name: {4-[(carbamothioylamino)carbamoyl]phenyl}boronic acid
Synonyms: 4-(carbamothioylaminocarbamoyl)phenylboronic acid
Molecular Weight (g/mol): 239.06
Molecular Formula: C8H10BN3O3S
InChi Key: NVXNXTLWXKMYHB-UHFFFAOYSA-N
SMILES: NC(=S)NNC(=O)C1=CC=C(C=C1)B(O)O

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results