accessibility menu, dialog, popup

UserName

CAS RN 95838-16-7

IUPAC Name: λ⁴-cerium(4+) hydrate disulfate
Synonyms: λ⁴-cerium(4+) hydrate disulfate
Molecular Weight (g/mol): 350.24
Molecular Formula: CeH2O9S2
InChi Key: KNANFSXYDCURQS-UHFFFAOYSA-J
SMILES: O.[Ce+4].[O-]S([O-])(=O)=O.[O-]S([O-])(=O)=O
Percent Purity 
  • (3)
  • (2)
Quantity 
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
Grade 
  • (2)
Physical Form 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (3)
  • (2)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (2)
  • (2)