accessibility menu, dialog, popup

UserName

CAS RN 96-82-2

HO(R)HO(R)O(S)O(R)HO(R)HO(S)OH(R)OH(R)OH(R)OHOHO

CAS RN 96-82-2

IUPAC Name: (2R,3R,4R,5R)-2,3,5,6-tetrahydroxy-4-{[(2S,3R,4S,5R,6R)-3,4,5-trihydroxy-6-(hydroxymethyl)oxan-2-yl]oxy}hexanoic acid
Synonyms: lactobionic acid
Molecular Weight (g/mol): 358.3
Molecular Formula: C12H22O12
InChi Key: JYTUSYBCFIZPBE-AMTLMPIISA-N
SMILES: OC[C@@H](O)[C@@H](O[C@@H]1O[C@H](CO)[C@H](O)[C@H](O)[C@H]1O)[C@H](O)[C@@H](O)C(O)=O
Percent Purity 
  • (2)
  • (3)
Physical Form 
  • (2)
  • (3)
Quantity 
  • (2)
  • (2)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (2)
  • (3)
  • (2)
  • (3)
  • (2)
  • (3)
  • (2)
  • (2)
  • (1)