accessibility menu, dialog, popup

UserName

CAS RN 96854-34-1

IUPAC Name: (1S)-1-phenylpropane-1,3-diol
Synonyms: (1S)-1-phenylpropane-1,3-diol
Molecular Weight (g/mol): 152.19
Molecular Formula: C9H12O2
InChi Key: RRVFYOSEKOTFOG-VIFPVBQESA-N
SMILES: OCC[C@H](O)C1=CC=CC=C1
Physical Form 
  • (3)
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results