CAS RN 96994-75-1
CAS RN 96994-75-1
IUPAC Name:
2-chloro-N,N-dimethyl-6-nitroaniline
Synonyms:
2-chloro-N,N-dimethyl-6-nitroaniline
Molecular Weight (g/mol):
200.62
Molecular Formula:
C8H9ClN2O2
InChi Key:
AMHCHWZVYGFSNK-UHFFFAOYSA-N
SMILES:
CN(C)C1=C(C=CC=C1Cl)[N+]([O-])=O
1
–
1
of
1
results
1
2-Chloro-N,N-dimethyl-6-nitroaniline, 96%, Thermo Scientific™
CAS: 96994-75-1 Molecular Formula: C8H9ClN2O2 Molecular Weight (g/mol): 200.622 InChI Key: AMHCHWZVYGFSNK-UHFFFAOYSA-N Synonym: 2-chloro-6-nitro-n,n-dimethylaniline,2-chloro-n,n-dimethyl-6-nitrobenzenamine,2-chloro-6-nitro-phenyl-dimethyl-amine PubChem CID: 13418267 IUPAC Name: 2-chloro-N,N-dimethyl-6-nitroaniline SMILES: CN(C)C1=C(C=CC=C1Cl)[N+](=O)[O-]