CAS RN 97744-44-0
CAS RN 97744-44-0
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
1
of
1
results
1
Dibenzylfluorescein, TRC
CAS: 97744-44-0 Molecular Formula: C34 H24 O5 Molecular Weight (g/mol): 512.551 Synonym: 2-[3-Oxo-6-(phenylmethoxy)-3H-xanthen-9-yl]benzoic Acid Phenylmethyl Ester,NSC 645658 IUPAC Name: 6,7-dibenzyl-3',6'-dihydroxyspiro[2-benzofuran-3,9'-xanthene]-1-one SMILES: Oc1ccc2c(Oc3cc(O)ccc3C24OC(=O)c5c(Cc6ccccc6)c(Cc7ccccc7)ccc45)c1