accessibility menu, dialog, popup

UserName

NaNaOONOON

CAS RN 979-88-4

IUPAC Name: disodium [2,2'-biquinoline]-4,4'-dicarboxylate
Synonyms: disodium [2,2'-biquinoline]-4,4'-dicarboxylate
Molecular Weight (g/mol): 388.29
Molecular Formula: C20H10N2Na2O4
InChi Key: AUPXFICLXPLHBB-UHFFFAOYSA-L
SMILES: [Na+].[Na+].[O-]C(=O)C1=C2C=CC=CC2=NC(=C1)C1=CC(C([O-])=O)=C2C=CC=CC2=N1
Molecular Weight (g/mol) 
  • (2)
  • (3)
Quantity 
  • (1)
  • (2)
  • (1)
  • (1)
Physical Form 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results