accessibility menu, dialog, popup

UserName

NOBr

CAS RN 97925-43-4

IUPAC Name: 4-bromo-1,2-oxazole
Synonyms: 4-bromo-1,2-oxazole
Molecular Weight (g/mol): 147.96
Molecular Formula: C3H2BrNO
InChi Key: IRDVIGFQMLUJAM-UHFFFAOYSA-N
SMILES: BrC1=CON=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results