accessibility menu, dialog, popup

UserName

CAS RN 98-60-2

IUPAC Name: 4-chlorobenzene-1-sulfonyl chloride
Synonyms: 4-chlorobenzenesulfonyl chloride
Molecular Weight (g/mol): 211.06
Molecular Formula: C6H4Cl2O2S
InChi Key: ZLYBFBAHAQEEQQ-UHFFFAOYSA-N
SMILES: ClC1=CC=C(C=C1)S(Cl)(=O)=O
Molecular Weight (g/mol) 
  • (6)
  • (3)
Quantity 
  • (1)
  • (1)
  • (2)
  • (1)
  • (2)
  • (2)
Boiling Point 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (4)
  • (2)
  • (3)
  • (6)
  • (3)
  • (1)
  • (1)
  • (2)
  • (1)
  • (2)
  • (2)
  • (4)
  • (2)