accessibility menu, dialog, popup

UserName

CAS RN 98-89-5

HOO

CAS RN 98-89-5

IUPAC Name: cyclohexanecarboxylic acid
Synonyms: cyclohexanecarboxylic acid calcium salt cyclohexanecarboxylic acid cobalt salt cyclohexanecarboxylic acid epa pesticide chemical code 112603 potassium salt cyclohexanecarboxylic acid sodium salt cyclohexanecarboxylic acid lead salt cyclohexanecarboxylic acid cyclohexancarbonsaeure
Molecular Weight (g/mol): 128.17
Molecular Formula: C7H12O2
InChi Key: NZNMSOFKMUBTKW-UHFFFAOYSA-N
SMILES: OC(=O)C1CCCCC1
Molecular Weight (g/mol) 
  • (7)
  • (3)
Quantity 
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (2)
  • (1)
Boiling Point 
  • (3)
  • (4)
Physical Form 
  • (4)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (4)
  • (3)
  • (3)
  • (7)
  • (3)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (2)
  • (1)
  • (3)
  • (4)
  • (4)