accessibility menu, dialog, popup

UserName

H3CNH3CPNCH3H3CCH3NNH3CCH3

CAS RN 98015-45-3

IUPAC Name: 2-(tert-butylimino)-N,N-diethyl-1,3-dimethyl-1,3,2λ⁵-diazaphosphinan-2-amine
Synonyms: 2-(tert-butylimino)-N,N-diethyl-1,3-dimethyl-1,3,2λ⁵-diazaphosphinan-2-amine
Molecular Weight (g/mol): 274.39
Molecular Formula: C13H31N4P
InChi Key: VSCBATMPTLKTOV-UHFFFAOYSA-N
SMILES: CCN(CC)P1(=NC(C)(C)C)N(C)CCCN1C
Boiling Point 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results