accessibility menu, dialog, popup

UserName

CAS RN 98700-50-6

IUPAC Name: 5-methyl-1-phenyl-1H-pyrazole-4-carbaldehyde
Synonyms: 5-methyl-1-phenylpyrazole-4-carbaldehyde
Molecular Weight (g/mol): 186.21
Molecular Formula: C11H10N2O
InChi Key: IIEFFVFWBUGUTI-UHFFFAOYSA-N
SMILES: CC1=C(C=O)C=NN1C1=CC=CC=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results