accessibility menu, dialog, popup

UserName

CH3NHCH3NOOFFFOOH3C

CAS RN 98791-67-4

IUPAC Name: methyl 2,6-dimethyl-5-nitro-4-[2-(trifluoromethyl)phenyl]-1,4-dihydropyridine-3-carboxylate
Synonyms: methyl 2,6-dimethyl-5-nitro-4-[2-(trifluoromethyl)phenyl]-1,4-dihydropyridine-3-carboxylate
Molecular Weight (g/mol): 356.3
Molecular Formula: C16H15F3N2O4
InChi Key: ZFLWDHHVRRZMEI-UHFFFAOYNA-N
SMILES: COC(=O)C1=C(C)NC(C)=C(C1C1=CC=CC=C1C(F)(F)F)[N+]([O-])=O
Grade 
  • (3)
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results