accessibility menu, dialog, popup

UserName

H3COONH2HCl

CAS RN 98959-77-4

IUPAC Name: 2-(4-methoxyphenoxy)ethan-1-amine hydrochloride
Synonyms: 2-(4-methoxyphenoxy)ethanamine hydrochloride
Molecular Weight (g/mol): 203.67
Molecular Formula: C9H14ClNO2
InChi Key: YCFXTJDWVNEVEX-UHFFFAOYSA-N
SMILES: Cl.COC1=CC=C(OCCN)C=C1
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results