accessibility menu, dialog, popup

UserName

CAS RN 99474-02-9

H3COBr

CAS RN 99474-02-9

IUPAC Name: 1-(4-bromophenyl)heptan-1-one
Synonyms: 1-(4-bromophenyl)heptan-1-one
Molecular Weight (g/mol): 269.18
Molecular Formula: C13H17BrO
InChi Key: LDVBFPQTLJKTLV-UHFFFAOYSA-N
SMILES: CCCCCCC(=O)C1=CC=C(Br)C=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results