accessibility menu, dialog, popup

UserName

CH3OPOPOOH3COH3COH3C

CAS RN 995-32-4

IUPAC Name: diethyl [2-(diethoxyphosphoryl)ethyl]phosphonate
Synonyms: diethyl 2-(diethoxyphosphoryl)ethylphosphonate
Molecular Weight (g/mol): 302.24
Molecular Formula: C10H24O6P2
InChi Key: RSQYXXACEZCDFS-UHFFFAOYSA-N
SMILES: CCOP(=O)(CCP(=O)(OCC)OCC)OCC
Quantity 
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
Boiling Point 
  • (3)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (3)
  • (3)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (3)