accessibility menu, dialog, popup

UserName

OH(R)CHFFF

CAS RN 99727-20-5

IUPAC Name: (2R)-1,1,1-trifluoro-2-phenylbut-3-yn-2-ol
Synonyms: (2R)-1,1,1-trifluoro-2-phenylbut-3-yn-2-ol
Molecular Weight (g/mol): 200.16
Molecular Formula: C10H7F3O
InChi Key: WWIJKFJKIOQDKI-SECBINFHSA-N
SMILES: O[C@](C#C)(C1=CC=CC=C1)C(F)(F)F

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results