accessibility menu, dialog, popup

UserName

CH3GeCH3CH3

CAS RN 998-39-0

IUPAC Name: tributylgermyl
Synonyms: tributylgermyl
Molecular Weight (g/mol): 243.98
Molecular Formula: C12H27Ge
InChi Key: UMRPCNGKANTZSN-UHFFFAOYSA-N
SMILES: CCCC[Ge](CCCC)CCCC
Boiling Point 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results