UserName
Five Things to Consider When Buying Analytical Balances
Featured Products
Analytical Balances
An analytical balance is a sensitive piece of lab equipment that enables precise measurement of mass due to its high readability and draught proof weighing chamber. With an analytical balance it's possible to measure to a precision of 0.0001g, making them the ideal choice for pharmaceutical, industrial, chemical, and food and beverage laboratories.
They usually have lower capacity limits compared to other types of balances, often ranging from 100g to 200g.
Some may have internal calibration features while others requires external calibration.
For both weighing and calibration, analytical balances should be placed on an anti-vibration table as they can sense the smallest variation in mass.
Features like removable draft shields and spill-resistant designs can simplify upkeep and prolong the balance's lifespan.
Popular Resources
Top Tips for Lab Weighing with OHAUS
External factors such as temperature fluctuations, air currents, electromagnetic interference, and vibrations can all have an impact on the accuracy of an analytical balance.
28392-analytical-balances-1
Cubis II Ultra-High Resolution Balances
The Cubis II MCA66S showed excellent repeatability values with all three different types of vessels. The measurement time is comparable or slightly better in contrast to the tested competitor.
28392-analytical-balances-2
A&D Advanced-Level Analytical/Precision Balances
All the innovative technologies built into the A&D Apollo are designed to provide functional solutions to the real world problems that users face in laboratories or in factories.
28392-analytical-balances-3
Five Things to Consider When Buying Balances and Scales
Different scales have varying capacities, from a few grams for precision scales to several tons for industrial scales. Ensure the scale's capacity and readability align with your requirements.
28392-image2
Top Brands