missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Loperamide-d6 HCl (dimethyl-d6), CDN
Click to view available options
Quantity:
0.01 g
0.05 g
Unit Size:
0.01g
0.05g
Specifications
Specifications
| Chemical Name or Material | Loperamide-d6 Hydrochloride |
| CAS | 1189469-46-2 |
| Molecular Formula | C29 D6 H27 Cl N2 O2 . Cl H |
| Quantity | 0.05 g |
| Synonym | 4-[4-(4-Chlorophenyl)-4-hydroxypiperidin-1-yl]-2,2-diphenyl-N,N-bis(trideuteriomethyl)butanamide hydrochloride, 4-(4-Chlorophenyl)-4-hydroxy-N,N-di(methyl-d3)-α,α-diphenyl-1-piperidinebutanamide hydrochloride (1:1), Loperamide-d6 hydrochloride, Loperamide-D6 (N,N-dimethyl-D6) Hydrochloride, Loperamide-d6 HCl (dimethyl-d6) |
| InChI Formula | InChI=1 S/C29H33ClN2O2.ClH/c1-31(2)27(33)29(24-9-5-3-6-10-24,25-11-7-4-8-12-25)19-22-32-20-17-28(34,18-21-32)23-13-15-26(30)16-14-23;/h3-16,34 H,17-22H2,1-2H3;1 H/i1D3,2D3; |
| SMILES | Cl.[2 H]C([2 H])([2 H])N(C(=O)C(CCN1CCC(O)(CC1)c2ccc(Cl)cc2)(c3ccccc3)c4ccccc4)C([2 H])([2 H])[2 H] |
| IUPAC Name | 4-[4-(4-chlorophenyl)-4-hydroxypiperidin-1-yl]-2,2-diphenyl-N,N-bis(trideuteriomethyl)butanamide;hydrochloride |
| Molecular Weight (g/mol) | 519.5354 |
| Formula Weight | 518.2374 g/mol |
| Show More |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction