missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Methyl Heptadecanoate-d33, CDN
Click to view available options
Quantity:
0.1 g
0.5 g
Unit Size:
0.1g
0.50g
Specifications
Specifications
| Chemical Name or Material | Methyl Margarate-d33 |
| CAS | 1219804-81-5 |
| Molecular Formula | C18 2H33 H3 O2 |
| Quantity | 0.1 g |
| InChI Formula | InChI=1 S/C18H36O2/c1-3-4-5-6-7-8-9-10-11-12-13-14-15-16-17-18(19)20-2/h3-17H2,1-2H3/i1D3,3D2,4D2,5D2,6D2,7D2,8D2,9D2,10D2,11D2,12D2,13D2,14D2,15D2,16D2,17D2 |
| SMILES | [2 H]C([2 H])([2 H])C([2 H])([2 H])C([2 H])([2 H])C([2 H])([2 H])C([2 H])([2 H])C([2 H])([2 H])C([2 H])([2 H])C([2 H])([2 H])C([2 H])([2 H])C([2 H])([2 H])C([2 H])([2 H])C([2 H])([2 H])C([2 H])([2 H])C([2 H])([2 H])C([2 H])([2 H])C([2 H])([2 H])C(=O)OC |
| IUPAC Name | methyl 2,2,3,3,4,4,5,5,6,6,7,7,8,8,9,9,10,10,11,11,12,12,13,13,14,14,15,15,16,16,17,17,17-tritriacontadeuterioheptadecanoate |
| Molecular Weight (g/mol) | 317.68 |
| Formula Weight | 317.4787 g/mol |
| Percent Purity | 98 atom % D, min 98% Chemical Purity |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction