Learn More
Trajan Scientific and Medical™ 0.36mm PEEKsil™ Tubing
A mechanically robust polymer-sheathed fused silica tubing for use in HPLC systems
43.00 € - 77.30 €
Product Code | Brand | Diameter (Metric) Inner | Overall Length (Metric) | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Diameter (Metric) Inner | Overall Length (Metric) | Price | Quantity & Availability | ||||
15288783
|
Trajan Scientific and Medical™
0624371 |
0.025 mm | 50 m |
43.00 €
Pack of 2 |
Estimated Shipment: 15-05-2024
Log in to see stock. |
||||
15298783
|
Trajan Scientific and Medical™
0624372 |
0.025 mm | 100 m |
46.30 €
Pack of 2 |
Estimated Shipment: 15-05-2024
Log in to see stock. |
||||
15208793
|
Trajan Scientific and Medical™
0624373 |
0.025 mm | 150 m |
49.50 €
Pack of 2 |
Estimated Shipment: 15-05-2024
Log in to see stock. |
||||
15218793
|
Trajan Scientific and Medical™
0624374 |
0.025 mm | 250 m |
55.90 €
Pack of 2 |
Estimated Shipment: 13-06-2024
Log in to see stock. |
||||
15228793
|
Trajan Scientific and Medical™
0624375 |
0.025 mm | 500 m |
77.30 €
Pack of 2 |
Estimated Shipment: 13-06-2024
Log in to see stock. |
||||
15238793
|
Trajan Scientific and Medical™
0624376 |
0.05 mm | 50 m |
43.00 €
Pack of 2 |
Estimated Shipment: 15-05-2024
Log in to see stock. |
||||
15248793
|
Trajan Scientific and Medical™
0624377 |
0.05 mm | 100 m |
46.30 €
Pack of 2 |
Estimated Shipment: 15-05-2024
Log in to see stock. |
||||
15258793
|
Trajan Scientific and Medical™
0624378 |
0.05 mm | 150 m |
49.50 €
Pack of 2 |
Estimated Shipment: 15-05-2024
Log in to see stock. |
||||
15268793
|
Trajan Scientific and Medical™
0624379 |
0.05 mm | 250 m |
55.90 €
Pack of 2 |
Estimated Shipment: 15-05-2024
Log in to see stock. |
||||
15278793
|
Trajan Scientific and Medical™
0624380 |
0.05 mm | 500 m |
77.30 €
Pack of 2 |
Estimated Shipment: 15-05-2024
Log in to see stock. |
||||
Description
- Fused silica internal wall provides smooth surface, free of imperfections
- PEEKsil compatible with most organic solvents and resistant to strong acids
- Effective pH range of 0 to 10
- Flexibility ensures easy connection between columns, detectors and injectors
Specifications
Tubing | |
2/Pk. |