missing translation for 'onlineSavingsMsg'
Learn More

1,1-Di-(tert-butylperoxy)-3,3,5-trimethylcyclohexane, 75% solution in aromatic free mineral spirit, Thermo Scientific Chemicals

 

Brand:  Thermo Scientific Acros 349772500

Code : 29

Additional Details : CAS Number : 6731-36-8 Weight : 0.21750kg Transport : UN number : 3103 Chem class : 5.2


Product Code. 10595671

  • 84.80 € / 250mL
In Stock
to see stock.

Please to purchase this item. Need a web account? Register with us today!


Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Radical initiators
Specifications

Specifications

1,1-Di-(tert-butylperoxy)-3,3,5-trimethylcyclohexane
75% Sol.in Arom.free Min.spirit
24.0
C2H4Cl2
MFCD00084801
ethylidene chloride, ethane, 1,1-dichloro, ethylidene dichloride, dichloroethane, 1,1-dichlorethane, aethylidenchlorid, 1,1-dicloroetano, 1,1-ethylidene dichloride, dichloromethylmethane, rcra waste number u076
CC(Cl)Cl
98.954
CHEBI:81599
74 to 76%
250 mL
Undesignated
75-34-3,67-56-1
26.0
[(CH3)3COO]2C6H7(CH3)3
0.8950g/mL
SCYULBFZEHDVBN-UHFFFAOYSA-N
1,1-dichloroethane
6365
302.45
Plastic bottle
Solution
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

Product Identifier
  • 1,1-Di-(tert-butylperoxy)-3,3,5-trimethylcyclohexane,75% sol.in arom.free min.spirit

Signal Word
  • Danger

Hazard Category
  • Organic peroxide Type C & D

Hazard Statement
  • H242-Heating may cause a fire.

Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P220-Keep/Store away from clothing/combustible materials.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.

SDS Documents

Documents

Certificates
Special Offers

Special Offers

RUO – Research Use Only