missing translation for 'onlineSavingsMsg'
Learn More

1,1'-Thiocarbonyldiimidazole, tech 90%, Thermo Scientific Chemicals

Catalog Number p-7026167
Click to view available options
Quantity:
10 g
50 g
250 g
This item is not returnable. View return policy
6160-65-2
C7H6N4S
178.21
MFCD00005289
RAFNCPHFRHZCPS-UHFFFAOYSA-N
1,1'-thiocarbonyldiimidazole, thiocarbonyldiimidazole, n,n'-thiocarbonyldiimidazole, di 1h-imidazol-1-yl methanethione, 1,1-thiocarbonyldiimidazole, di imidazol-1-yl methanethione, n,n'-thiocarbonyldiimidazole tcdi, 1h-imidazole, 1,1'-carbonothioylbis, n,n-thiocarbonyldiimidazole, 1,1'-carbonothioylbis-1h-imidazole
80264
di(imidazol-1-yl)methanethione
S=C(N1C=CN=C1)N1C=CN=C1
This item is not returnable. View return policy
6160-65-2
C7H6N4S
178.21
MFCD00005289
RAFNCPHFRHZCPS-UHFFFAOYSA-N
1,1'-thiocarbonyldiimidazole, thiocarbonyldiimidazole, n,n'-thiocarbonyldiimidazole, di 1h-imidazol-1-yl methanethione, 1,1-thiocarbonyldiimidazole, di imidazol-1-yl methanethione, n,n'-thiocarbonyldiimidazole tcdi, 1h-imidazole, 1,1'-carbonothioylbis, n,n-thiocarbonyldiimidazole, 1,1'-carbonothioylbis-1h-imidazole
80264
di(imidazol-1-yl)methanethione
S=C(N1C=CN=C1)N1C=CN=C1

1,1'-Thiocarbonyldiimidazole is used as a reagent for the deoxygenation of alcohols with tributyltin hydride and diols with lithium aluminum hydride. It is also used to deoxygenate carboxylic monosaccharide analogues. Further, it is a useful reagent involved in biochemical synthesis. In addition to this, it is used to prepare 1-[bis-(4-methoxy-phenyl)-methyl]-1H-imidazole by reacting with 4,4'-dimethoxy-benzhydrol.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
1,1′-Thiocarbonyldiimidazole is used as a reagent for the deoxygenation of alcohols with tributyltin hydride and diols with lithium aluminum hydride. It is also used to deoxygenate carboxylic monosaccharide analogues. Further, it is a useful reagent involved in biochemical synthesis. In addition to this, it is used to prepare 1-[bis-(4-methoxy-phenyl)-methyl]-1H-imidazole by reacting with 4,4′-dimethoxy-benzhydrol.

Solubility
Soluble in terahydrofuran, toluene and dichloromethane.

Notes
Air and moisture sensitive. Keep the container tightly closed in a dry and well-ventilated place. Store in a cool place. Incompatible with strong oxidizing agents and strong acids.
TRUSTED_SUSTAINABILITY
Melting Point 98°C to 102°C
Assay Percent Range 0.95
Quantity 10 g
Beilstein 609349
Sensitivity Moisture sensitive
Solubility Information Soluble in terahydrofuran,toluene and dichloromethane.
Formula Weight 178.22
Percent Purity 90%
Grade Technical
Chemical Name or Material 1, 1'-Thiocarbonyldiimidazole

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'