missing translation for 'onlineSavingsMsg'
Learn More

1,10-Phenanthroline monohydrate, ACS reagent, Thermo Scientific Chemicals

Catalog Number p-180506
Click to view available options
Quantity:
1 g
5 g
25 g
100 g
Packaging:
Glass bottle
This item is not returnable. View return policy
5144-89-8, 66-71-7
C12H10N2O
198.23
MFCD00149973
PPQJCISYYXZCAE-UHFFFAOYSA-N
21226
O.C1=CN=C2C(C=CC3=CC=CN=C23)=C1
This item is not returnable. View return policy
5144-89-8, 66-71-7
C12H10N2O
198.23
MFCD00149973
PPQJCISYYXZCAE-UHFFFAOYSA-N
21226
O.C1=CN=C2C(C=CC3=CC=CN=C23)=C1

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Indicator
TRUSTED_SUSTAINABILITY
Melting Point 97°C to 101°C
Color White
Infrared Spectrum Authentic
Packaging Glass bottle
Quantity 1 g
Beilstein 23, 227
Merck Index 15, 7328
Synonym 1,10-phenanthroline hydrate, 1,10-phenanthroline monohydrate, o-phenanthroline monohydrate, unii-ksx215x00e, 1,10-fenantrolina, 4,5-phenanthroline monohydrate, 1,10-phenanthroline, monohydrate, 1,10-fenanthrolin, 1,10-phenanthrolineo-phenanthroline, 1, 10-phenanthroline monohydrate
Solubility Information Solubility in water: slightly soluble. Other solubilities: soluble in alcohol,acetone and benzene
IUPAC Name 1,10-phenanthroline;hydrate
Formula Weight 198.23
Grade ACS Reagent
Physical Form Needle-like Crystals
Chemical Name or Material 1, 10-Phenanthroline monohydrate
Show More Show Less

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'