missing translation for 'onlineSavingsMsg'
Learn More

1-[2-(2,4-Difluorophenyl)-2,3-epoxypropyl]-1H-1,2,4-triazole methanesulfonate, 98%, Thermo Scientific Chemicals

Catalog Number p-7053975
Click to view available options
Quantity:
1 g
5 g
This item is not returnable. View return policy
86386-77-8
MFCD11111978
1-[2-(2,4-difluorophenyl)-2-oxiranylmethyl]-1H-1,2,4-triazole methanesulfonate
This item is not returnable. View return policy
86386-77-8
MFCD11111978
1-[2-(2,4-difluorophenyl)-2-oxiranylmethyl]-1H-1,2,4-triazole methanesulfonate

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
Quantity 1 g
Formula Weight 333.31
Percent Purity 98%
Physical Form Solid
Chemical Name or Material 1-[2-(2,4-Difluorophenyl)-2,3-epoxypropyl]-1H-1,2,4-triazole methanesulfonate

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'