missing translation for 'onlineSavingsMsg'
Learn More

Alfa Aesar™ 1,2,3,4-Tetrahydro-2-naphthylamine, 97%

Brand:  Alfa Aesar™ H66652.06

Code : NEW

Additional Details : Weight : 0.00500kg

Quantity 5g

 View more versions of this product

Product Code. 15472799

  • 984.30 € / 5g


Estimated Shipment date
from Supplier 20-06-2019
Add to basket

Specifications

Chemical Identifiers

2954-50-9
147.221
LCGFVWKNXLRFIF-UHFFFAOYSA-N
34677
C1CC2=CC=CC=C2CC1N
C10H13N
MFCD00045577
1,2,3,4-tetrahydro-2-naphthylamine, 2-aminotetralin, 2-naphthalenamine, 1,2,3,4-tetrahydro, 1,2,3,4-tetrahydro-2-naphthalenamine, 1,2,3,4-tetrahydro-naphthalen-2-ylamine, chembl30294, 2-1,2,3,4-tetrahydronaphthylamine, 2-amino-1,2,3,4-tetrahydronaphthalene, 2-amino-tetralin, tetralin-2-amine
1,2,3,4-tetrahydronaphthalen-2-amine

Specifications

1,2,3,4-Tetrahydro-2-naphthylamine
2954-50-9
MFCD00045577
1,2,3,4-tetrahydro-2-naphthylamine, 2-aminotetralin, 2-naphthalenamine, 1,2,3,4-tetrahydro, 1,2,3,4-tetrahydro-2-naphthalenamine, 1,2,3,4-tetrahydro-naphthalen-2-ylamine, chembl30294, 2-1,2,3,4-tetrahydronaphthylamine, 2-amino-1,2,3,4-tetrahydronaphthalene, 2-amino-tetralin, tetralin-2-amine
C1CC2=CC=CC=C2CC1N
147.221
147.1
97%
5g
C10H13N
1102357
LCGFVWKNXLRFIF-UHFFFAOYSA-N
1,2,3,4-tetrahydronaphthalen-2-amine
34677
Solid
Videos
SDS