missing translation for 'onlineSavingsMsg'
Learn More
Learn More
1-(2-Aminoethyl)-2-methyl-5-nitroimidazole dihydrochloride monohydrate, 98%, Thermo Scientific Chemicals
Click to view available options
5g; Glass bottle
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
CAS | 49575-10-2 |
CAS Min % | 97.5 |
CAS Max % | 100.0 |
Melting Point | 195°C |
Assay Percent Range | 98% |
Packaging | Glass bottle |
Molecular Formula | 2HCl·H2O |
MDL Number | MFCD00066965 |
Quantity | 5 g |
Synonym | dihydrogen 2-2-methyl-5-nitroimidazol-1-yl ethanamine hydrate dichloride |
Show More |
Safety and Handling
Product Identifier
- 1-(2-Aminoethyl)-2-methyl-5-nitroimidazole dihydrochloride monohydrate
- Warning
- Acute toxicity Category 4
- Carcinogenicity Category 2
- H351-Suspected of causing cancer.
- H302-Harmful if swallowed.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P301+P312-IF SWALLOWED: Call a POISON CENTER or doctor/physician/ if you feel unwell.
RUO – Research Use Only
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'