missing translation for 'onlineSavingsMsg'
Learn More

1,2-Bis(4-pyridyl)ethane, 97%, Thermo Scientific Chemicals

Catalog Number p-3756936
Click to view available options
Quantity:
1 g
10 g
Packaging:
Glass bottle
This item is not returnable. View return policy
4916-57-8
C12H12N2
184.24
MFCD00006451
DQRKTVIJNCVZAX-UHFFFAOYSA-N
1,2-bis 4-pyridyl ethane, 1,2-di pyridin-4-yl ethane, 4,4'-ethylenedipyridine, 1,2-di 4-pyridyl ethane, 4,4'-ethane-1,2-diyldipyridine, unii-wo1evc302b, pyridine, 4,4'-1,2-ethanediyl bis, wo1evc302b, chembl70083, 4-2-pyridin-4-ylethyl pyridine
78630
4-(2-pyridin-4-ylethyl)pyridine
C1=CN=CC=C1CCC2=CC=NC=C2
This item is not returnable. View return policy
4916-57-8
C12H12N2
184.24
MFCD00006451
DQRKTVIJNCVZAX-UHFFFAOYSA-N
1,2-bis 4-pyridyl ethane, 1,2-di pyridin-4-yl ethane, 4,4'-ethylenedipyridine, 1,2-di 4-pyridyl ethane, 4,4'-ethane-1,2-diyldipyridine, unii-wo1evc302b, pyridine, 4,4'-1,2-ethanediyl bis, wo1evc302b, chembl70083, 4-2-pyridin-4-ylethyl pyridine
78630
4-(2-pyridin-4-ylethyl)pyridine
C1=CN=CC=C1CCC2=CC=NC=C2

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
CAS Min % 96.0
CAS Max % 100.0
Melting Point 107.0°C to 113.0°C
Color Pink or White to Yellow
Infrared Spectrum Authentic
Assay Percent Range 97%
Packaging Glass bottle
Linear Formula (C5H4N)CH2CH2(C5H4N)
Quantity 1 g
Formula Weight 184.24
Percent Purity 97%
Physical Form Crystalline Powder or Crystals
Chemical Name or Material 1, 2-Bis(4-pyridyl)ethane
Show More Show Less
compliance-icons
Product Identifier
  • 1,2-Bis(4-pyridyl)ethane
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H315-Causes skin irritation.
Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P264-Wash thoroughly after handling.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'