missing translation for 'onlineSavingsMsg'
Learn More

1,2-Bis(diphenylphosphino)ethane nickel(II) chloride, 99%, ACROS Organics™

Brand:  Acros Organics 363230010

Code : 29

Additional Details : CAS Number : 14647-23-5 Weight : 0.05100kg


Packaging Glass bottle
Quantity 1g

 View more versions of this product

Product Code. 10427022

  • 20.60 € / 1g


EU Stock 6
Add to basket


Specifications

Chemical Identifiers

1, 2-Bis(diphenylphosphino)ethane nickel(II) chloride
98.5
99%
MFCD00013313
14647-23-5
100.0
C26H26Cl2NiP2+2

Specifications

1, 2-Bis(diphenylphosphino)ethane nickel(II) chloride
14647-23-5
100.0
Authentic
Crystalline Powder
99%
MFCD00013313
263.0° to 265.0°C
C1=CC=C(C=C1)P(CCP(C2=CC=CC=C2)C3=CC=CC=C3)C4=CC=CC=C4.Cl.Cl.[Ni+2]
530.035
131664338
528.02
98.5
98.5% min.
Glass bottle
99%
C26H26Cl2NiP2+2
Orange to red
XXECWTBMGGXMKP-UHFFFAOYSA-N
2-diphenylphosphanylethyl(diphenyl)phosphane;nickel(2+);dihydrochloride
1g
Videos
Safety and Handling

Safety and Handling

Product Identifier
  • 1,2-Bis(diphenylphosphino)ethane nickel(II) chloride

Signal Word
  • Warning

Hazard Category
  • Carcinogenicity Category 2
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3

Hazard Statement
  • H319-Causes serious eye irritation.
  • H351-Suspected of causing cancer.
  • H335-May cause respiratory irritation.
  • H315-Causes skin irritation.

Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

SDS