Learn More
1-(2-Pyridylazo)-2-Naphthol, Pure, Fisher Chemical™
Brand: Fisher Chemical P/8000/44
Code : 21
Additional Details : CAS Number : 85-85-8 Weight : 0.00500kg
Packaging | Glass Jar |
---|---|
Quantity | 5g |
Chemical Identifiers
85-85-8 | |
249.273 | |
RAXUMGMWXZYADR-OBGWFSINSA-N | |
5376264 | |
C1=CC=C2C(=C1)C=CC(=O)C2=NNC3=CC=CC=N3 |
C15H11N3O | |
4071 | |
1-2-pyridylazo-2-naphthol, unii-095b53y3xv, pan indicator, 2-naphthalenol, 1-2-pyridinylazo, 1e-1-pyridin-2-ylhydrazinylidene naphthalen-2-one, 1-2-pyridylazo-2-hydroxynaphthalene, pan van, 1-2-pyridyldiazenyl naphthalen-2-ol, 1-pyridin-2-azo-2-naphthol, 2-naphthalenol, 1-2-2-pyridinyl diazenyl | |
(1E)-1-(pyridin-2-ylhydrazinylidene)naphthalen-2-one |
Specifications
85-85-8 | |
138°C | |
4071 | |
1-2-pyridylazo-2-naphthol, unii-095b53y3xv, pan indicator, 2-naphthalenol, 1-2-pyridinylazo, 1e-1-pyridin-2-ylhydrazinylidene naphthalen-2-one, 1-2-pyridylazo-2-hydroxynaphthalene, pan van, 1-2-pyridyldiazenyl naphthalen-2-ol, 1-pyridin-2-azo-2-naphthol, 2-naphthalenol, 1-2-2-pyridinyl diazenyl | |
RAXUMGMWXZYADR-OBGWFSINSA-N | |
(1E)-1-(pyridin-2-ylhydrazinylidene)naphthalen-2-one | |
5376264 | |
Powder Solid |
Red | |
C15H11N3O | |
Glass Jar | |
5g | |
C1=CC=C2C(=C1)C=CC(=O)C2=NNC3=CC=CC=N3 | |
249.273 | |
249.27g/mol |
Safety and Handling
- 1-(2-Pyridylazo)-2-naphthol
Signal Word
- Warning
Hazard Category
- Skin corrosion/irritation Category 2
- Serious eye damage/eye irritation Category 2
- Specific target organ toxicity Category 3
Hazard Statement
- H315-Causes skin irritation.
- H319-Causes serious eye irritation.
- H335-May cause respiratory irritation.
Precautionary Statement
- P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P302+P352-IF ON SKIN: Wash with plenty of water/soap.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.