Learn More
Sartorius™ 1.2. Standard Flexboy™ with EVAtubes
Brand:  Sartorius™ FFB102603
Additional Details : Weight : 0.75000kg
Product Code. 12996990
  • 672.04 € / Pack of 20
Estimated Shipment: 30-07-2024
to see stock.
Specifications
50 mL
Bags are designed for the preparation, storage and transport of biopharmaceutical solutions, intermediates and final bulk products
20 Pack
ISO11137
3 Ports
Gamma Irradiated